Gene Transcripts

Gene Symbol
Unc45a
Entrez Gene ID
101869
Gene Name
unc-45 homolog A (C. elegans)
Gene Aliases
AW538196
Chromosome Location
Chr. 7: 80325292 - 80341000 on Build GRCm38
UniGene ID
Mm.243024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133952.2 NP_598713.2 16 2238
GenBank mRNA AK028912.1 - 16 2224
AK047292.1 - 16 2185
AK047476.1 - 16 2185
AK088034.1 - 16 2159
AK166809.1 - 16 2221
BC004717.1 - 16 2227 3582116

View Full Product Details