Gene Transcripts

Gene Symbol
Plekho2
Entrez Gene ID
102595
Gene Name
pleckstrin homology domain containing, family O member 2
Gene Aliases
AI840980, Plekhq1
Chromosome Location
Chr. 9: 65552577 - 65580087 on Build GRCm38
UniGene ID
Mm.247303
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153119.3 NP_694759.1 3, 4 419
GenBank mRNA AK028069.1 - 3, 4 410
AK154092.1 - 3, 4 314
AK158288.1 - 3, 4 387
AK170479.1 - 3, 4 403
BC028908.1 - 3, 4 378 3668775

View Full Product Details