Gene Transcripts

Gene Symbol
Dhx57
Entrez Gene ID
106794
Gene Name
DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57
Gene Aliases
AW494914
Chromosome Location
Chr. 17: 80238304 - 80290476 on Build GRCm38
UniGene ID
Mm.12412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163759.1 NP_001157231.1 5 1255
NM_198942.2 NP_945180.2 4 1096
GenBank mRNA AK053628.1 - 5 923
BC062952.1 - 4 1096 6853031
BC065169.1 - 5 922 6831314
BC066091.1 - 4 758 6841207

View Full Product Details