Gene Transcripts

Gene Symbol
Afp
Entrez Gene ID
11576
Gene Name
alpha fetoprotein
Gene Aliases
Chromosome Location
Chr. 5: 90490714 - 90508907 on Build GRCm38
UniGene ID
Mm.358570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007423.4 NP_031449.3 11, 12 1479
GenBank mRNA AK010934.1 - 11, 12 1455
AK075972.1 - 11, 12 1459
AK076053.1 - 11, 12 1455
AK076197.1 - 11, 12 1457
AK145975.1 - 11, 12 1455
AK146513.1 - 11, 12 1457
AK146524.1 - 11, 12 1457
AK167549.1 - 11, 12 1455
AK168426.1 - 11, 12 1458
AK168574.1 - 11, 12 1457
AK168576.1 - 11, 12 1455
AK168751.1 - 11, 12 1457
AK168818.1 - 10, 11 1333
AK168883.1 - 11, 12 1455
AK169046.1 - 11, 12 1457
AK169054.1 - 11, 12 1457
AK169147.1 - 11, 12 1455
AV105248.2 - 2, 3 398
BC066206.1 - 11, 12 1442 30469872
V00743.1 - 11, 12 1453

View Full Product Details