Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007937.3 | NP_031963.2 | 3 | 1228 | |
GenBank mRNA | AK034289.1 | - | 3 | 1227 | |
AK049740.1 | - | 3 | 1229 | ||
AK089130.1 | - | 3 | 623 | ||
AK089143.1 | - | 3 | 1087 | ||
AK133698.1 | - | 3 | 1228 | ||
BC057401.1 | - | 3 | 1206 | 5697758 | |
U07357.1 | - | 3 | 768 |