Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003817.1 | NP_001003817.1 | 4 | 733 | |
GenBank mRNA | AK031099.1 | - | 4 | 601 | |
AK083669.1 | - | 4 | 601 | ||
AK129487.1 | - | 4 | 703 | ||
AK142296.1 | - | 4 | 726 | ||
BC046811.1 | - | 4 | 724 | 5702040 | |
BC053078.1 | - | 4 | 724 | 5702040 |