Gene Transcripts

Gene Symbol
Gdap2
Entrez Gene ID
14547
Gene Name
ganglioside-induced differentiation-associated-protein 2
Gene Aliases
C77050, D3Ertd801e
Chromosome Location
Chr. 3: 100162463 - 100206989 on Build GRCm38
UniGene ID
Mm.279228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010269.3 NP_034399.1 12, 13 1573
GenBank mRNA AK004887.1 - 12, 13 1519
AK082958.1 - 12, 13 1535
AK146835.1 - 12, 13 1563
AK161122.1 - 12, 13 1472
AK166799.1 - 12, 13 1488
AK166859.1 - 12, 13 1487
AK187786.1 - 4, 5 363
AK188572.1 - 3, 4 256
AK208171.1 - 3, 4 256
AK208831.1 - 3, 4 256
AK213851.1 - 3, 4 256
BC025070.1 - 12, 13 1482 3710712
Y17851.1 - 12, 13 1569

View Full Product Details