Gene Transcripts

Gene Symbol
Gdf5
Entrez Gene ID
14563
Gene Name
growth differentiation factor 5
Gene Aliases
BMP-14, Cdmp-1, bp, brp
Chromosome Location
Chr. 2: 155941023 - 155945364 on Build GRCm38
UniGene ID
Mm.4744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008109.2 NP_032135.2 1 843
GenBank mRNA AB259648.1 - 1 534
BC034546.1 - 1 855 4190036
U08337.1 - 1 855

View Full Product Details