Gene Transcripts

Gene Symbol
Hr
Entrez Gene ID
15460
Gene Name
hairless
Gene Aliases
ALUNC, AU, N, ba, bldy, hr, rh, rh-bmh, rhino
Chromosome Location
Chr. 14: 70554056 - 70573548 on Build GRCm38
UniGene ID
Mm.7598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021877.3 NP_068677.2 7 2472
GenBank mRNA AB221596.1 - 2 157
AB221597.1 - 5 1775
AY312855.1 - 6 2155
BC049182.1 - 7 2472 6468299

View Full Product Details