Gene Transcripts

Gene Symbol
Idh3g
Entrez Gene ID
15929
Gene Name
isocitrate dehydrogenase 3 (NAD+), gamma
Gene Aliases
Chromosome Location
Chr. X: 73778963 - 73786953 on Build GRCm38
UniGene ID
Mm.14825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008323.1 NP_032349.1 8 610
GenBank mRNA AJ748655.1 - 8 562
AK156309.1 - 8 608
AK166925.1 - 7 567
AK167131.1 - 5 1328
AK168526.1 - 8 608
BC085179.1 - 8 583 30699000
BC119170.1 - 8 593 8733919
BC119172.1 - 8 593 8733921
U68564.1 - 8 610

View Full Product Details