Gene Transcripts
- Gene Symbol
- Il15ra
- Entrez Gene ID
- 16169
- Gene Name
- interleukin 15 receptor, alpha chain
- Gene Aliases
- AA690181, IL-15RA
- Chromosome Location
- Chr. 2: 11705169 - 11733985 on Build GRCm38
- UniGene ID
- Mm.200196
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271497.1 |
NP_001258426.1
|
4, 5 |
646 |
|
NM_001271498.1 |
NP_001258427.1
|
3, 4 |
669 |
|
NM_001271499.1 |
NP_001258428.1
|
4, 5 |
610 |
|
NM_008358.2 |
NP_032384.1
|
4, 5 |
709 |
|
NM_133836.2 |
NP_598597.1
|
4, 5 |
762 |
|
GenBank mRNA |
AK040940.1 |
- |
3, 4 |
521 |
|
AK080000.1 |
- |
3, 4 |
521 |
|
AY219715.1 |
- |
4, 5 |
511 |
|
AY221616.1 |
- |
4, 5 |
387 |
|
AY221619.1 |
- |
3, 4 |
408 |
|
BC022705.1 |
- |
4, 5 |
519 |
|
BC095982.1 |
- |
1, 2 |
437 |
|
BC132231.1 |
- |
3, 4 |
309 |
|
BC132233.1 |
- |
3, 4 |
309 |
|
BC145348.1 |
- |
3, 4 |
309 |
|
View Full Product Details