Gene Transcripts

Gene Symbol
Irs3
Entrez Gene ID
16369
Gene Name
insulin receptor substrate 3
Gene Aliases
AA408304
Chromosome Location
Chr. 5: 137643030 - 137645748 on Build GRCm38
UniGene ID
Mm.24486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010571.3 NP_034701.1 2 1090
GenBank mRNA AF035948.1 - 2 896
AK030311.1 - 2 1092
AK040947.1 - 2 1075
BC098235.1 - 2 922 1546952

View Full Product Details