Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181325.4 | NP_851842.1 | 6 | 882 | |
GenBank mRNA | AF133914.1 | - | 6 | 730 | |
AK052758.1 | - | 6 | 814 | ||
AK081243.1 | - | 5 | 3006 | ||
AK142223.1 | - | 6 | 752 | ||
AK145966.1 | - | 6 | 752 | ||
AK154988.1 | - | 6 | 859 | ||
BC061143.1 | - | 6 | 523 | 30276598 |