Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013628.2 | NP_038656.1 | 1 | 163 | |
GenBank mRNA | AK043742.1 | - | 1 | 235 | |
BC108982.1 | - | 1 | 141 | 40047652 | |
BC108983.1 | - | 1 | 141 | 40047655 | |
M58589.1 | - | 1 | 163 | ||
M69196.1 | - | 1 | 269 | ||
X57088.1 | - | 1 | 275 |