Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011081.2 | NP_035211.2 | 5 | 1137 | |
GenBank mRNA | AK014488.1 | - | 3 | 2725 | |
AK032283.1 | - | 5 | 1108 | ||
AK161026.1 | - | 5 | 1107 | ||
AK171453.1 | - | 5 | 730 | ||
BC035538.1 | - | 4 | 798 | ||
BC051166.1 | - | 4 | 358 | ||
BC058767.1 | - | 3 | 245 | ||
BC138755.1 | - | 4 | 1095 | 8861777 | |
BC138758.1 | - | 4 | 1095 | 8861780 | |
D26047.1 | - | 5 | 1106 | ||
S78188.1 | - | 5 | 1143 |