Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008846.2 | NP_032872.1 | 8 | 1053 | |
GenBank mRNA | AK133809.1 | - | 8 | 1048 | |
AK146100.1 | - | 8 | 1053 | ||
AK149354.1 | - | 8 | 1053 | ||
AK164552.1 | - | 8 | 1036 | ||
BC034864.1 | - | 8 | 1067 | 3326897 | |
D86176.1 | - | 7 | 986 |