Gene Transcripts

Gene Symbol
Ppt1
Entrez Gene ID
19063
Gene Name
palmitoyl-protein thioesterase 1
Gene Aliases
9530043G02Rik, AA960502, C77813, CLN1, D4Ertd184e, INCL, PPT
Chromosome Location
Chr. 4: 122836075 - 122859175 on Build GRCm38
UniGene ID
Mm.277719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008917.3 NP_032943.2 5 530
GenBank mRNA AF071025.1 - 5 547
AF087568.1 - 5 508
AF326557.1 - 5 514
AF326558.1 - 5 514
AK014561.1 - 5 512
AK018046.1 - 1 1008
AK148314.1 - 5 505
AK151055.1 - 5 505
AK151107.1 - 5 516
AK152474.1 - 5 514
AK152940.1 - 5 516
AK153106.1 - 5 511
AK153316.1 - 5 511
AK169579.1 - 6 605
AK169658.1 - 6 611
AK171668.1 - 5 512
AK216167.1 - 3 182

View Full Product Details