Gene Transcripts
- Gene Symbol
- Slc12a2
- Entrez Gene ID
- 20496
- Gene Name
- solute carrier family 12, member 2
- Gene Aliases
- 9330166H04Rik, Nkcc1, mBSC2, sy-ns
- Chromosome Location
- Chr. 18: 57878513 - 57946821 on Build GRCm38
- UniGene ID
- Mm.399997
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009194.3 |
NP_033220.2
|
12 |
2026 |
|
GenBank mRNA |
U13174.1 |
- |
12 |
2023 |
|
View Full Product Details