Gene Transcripts
- Gene Symbol
- Smpd1
- Entrez Gene ID
- 20597
- Gene Name
- sphingomyelin phosphodiesterase 1, acid lysosomal
- Gene Aliases
- A-SMase, ASM, Zn-SMase, aSMase
- Chromosome Location
- Chr. 7: 105554360 - 105558389 on Build GRCm38
- UniGene ID
- Mm.4628
- Species
- Mus musculus
- Gene Symbol
- Eno1
- Entrez Gene ID
- 13806
- Gene Name
- enolase 1, alpha non-neuron
- Gene Aliases
- 0610008I15, AL022784, Eno-1, MBP-1
- Chromosome Location
- Chr. 4: 150236726 - 150248873 on Build GRCm38
- UniGene ID
- Mm.372357
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK193395.1 |
- |
|
278 |
|
View Full Product Details