Gene Transcripts
- Gene Symbol
- Baiap2l2
- Entrez Gene ID
- 207495
- Gene Name
- BAI1-associated protein 2-like 2
- Gene Aliases
- BC048937
- Chromosome Location
- Chr. 15: 79258195 - 79285532 on Build GRCm38
- UniGene ID
- Mm.309647
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177580.3 |
NP_808248.1
|
11 |
1348 |
|
GenBank mRNA |
BC048937.1 |
- |
11 |
1286 |
|
View Full Product Details