Gene Transcripts

Gene Symbol
Depdc7
Entrez Gene ID
211896
Gene Name
DEP domain containing 7
Gene Aliases
AV216087
Chromosome Location
Chr. 2: 104721784 - 104742801 on Build GRCm38
UniGene ID
Mm.288805
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144804.1 NP_659053.1 6 1121
GenBank mRNA AK134830.1 - 6 1141
AK144066.1 - 6 1134
AK162086.1 - 6 1145
AK162218.1 - 6 1144
AK165504.1 - 6 1134
BC013499.1 - 6 1121 4222591

View Full Product Details