Gene Transcripts

Gene Symbol
Traf5
Entrez Gene ID
22033
Gene Name
TNF receptor-associated factor 5
Gene Aliases
Chromosome Location
Chr. 1: 191997203 - 191999968 on Build GRCm38
UniGene ID
Mm.380491
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011633.2 NP_035763.2 1253
GenBank mRNA AK144694.1 - 1204
BC012702.1 - 1207 4218625
D78141.1 - 1230
D83528.1 - 1095

View Full Product Details