Gene Transcripts

Gene Symbol
Smgc
Entrez Gene ID
223809
Gene Name
submandibular gland protein C
Gene Aliases
2310010P21Rik, DXImx49e, Muc19, Sfc21
Chromosome Location
Chr. 15: 91838328 - 91861435 on Build GRCm38
UniGene ID
Mm.486225
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198927.3 NP_945121.1 5 597
GenBank mRNA AK132579.1 - 5 599
AY459348.1 - 5 598
BC055490.1 - 5 579
BC092073.1 - 4 418
BC138091.1 - 5 598 8861110

View Full Product Details