Gene Transcripts

Gene Symbol
Arhgap30
Entrez Gene ID
226652
Gene Name
Rho GTPase activating protein 30
Gene Aliases
6030405P05Rik, Gm102, mFLJ00267
Chromosome Location
Chr. 1: 171388960 - 171410240 on Build GRCm38
UniGene ID
Mm.251048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005508.2 NP_001005508.2 6 902
GenBank mRNA AK031319.1 - 6 673
AK147757.1 - 6 909
AK156351.1 - 6 891
AK220204.1 - 1 36
BC082573.1 - 6 850 30548035

View Full Product Details