Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
227059
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
2900042E17Rik, 5430433I10, mKIAA1265
Chromosome Location
Chr. 1: 46807544 - 46925772 on Build GRCm38
UniGene ID
Mm.233889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172653.2 NP_766241.2 2 89
GenBank mRNA BC052880.1 - 2 203
BC062918.1 - 2 89 6833098

View Full Product Details