Gene Transcripts

Gene Symbol
Zp1
Entrez Gene ID
22786
Gene Name
zona pellucida glycoprotein 1
Gene Aliases
Chromosome Location
Chr. 19: 10914296 - 10920601 on Build GRCm38
UniGene ID
Mm.24767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009580.2 NP_033606.2 7 1136
GenBank mRNA BC100347.1 - 7 1154
BC125613.1 - 7 1136 40130128
BC125615.1 - 7 1136 40130130
U20448.1 - 7 1167

View Full Product Details