Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172685.3 | NP_766273.1 | 4, 5 | 555 | |
GenBank mRNA | AK032806.1 | - | 4, 5 | 556 | |
AK146034.1 | - | 4, 5 | 554 | ||
AK167778.1 | - | 4, 5 | 546 | ||
AK168090.1 | - | 4, 5 | 546 | ||
AK170598.1 | - | 4, 5 | 547 | ||
BC055369.1 | - | 4, 5 | 544 | 30055307 |