Gene Transcripts

Gene Symbol
Il20ra
Entrez Gene ID
237313
Gene Name
interleukin 20 receptor, alpha
Gene Aliases
E230031K19
Chromosome Location
Chr. 10: 19711673 - 19760053 on Build GRCm38
UniGene ID
Mm.234667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172786.2 NP_766374.1 7 1456
GenBank mRNA AK054215.1 - 7 1457
BC056628.1 - 7 1440 5033177

View Full Product Details