Gene Transcripts

Gene Symbol
Smcr8
Entrez Gene ID
237782
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8 homolog (human)
Gene Aliases
2310076G09Rik, AI642055, D030073L15Rik
Chromosome Location
Chr. 11: 60777525 - 60788287 on Build GRCm38
UniGene ID
Mm.57726
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085440.1 NP_001078909.1 1 1875
NM_175491.4 NP_780700.1 1 1875
GenBank mRNA AK083739.1 - 1 1859
AK145010.1 - 1 1861
AK164045.1 - 1 1864
BC085095.1 - 1 1505 30732606

View Full Product Details