Gene Transcripts

Gene Symbol
Bves
Entrez Gene ID
23828
Gene Name
blood vessel epicardial substance
Gene Aliases
Pop1, Popdc1, mBVES
Chromosome Location
Chr. 10: 45335762 - 45372484 on Build GRCm38
UniGene ID
Mm.332793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024285.2 NP_077247.1 3 428
GenBank mRNA AF124510.1 - 2 289
AF204174.1 - 3 426
BC132018.1 - 2 318 40130295
BC132044.1 - 2 318 40130321

View Full Product Details