Gene Transcripts
- Gene Symbol
- Nlrp5
- Entrez Gene ID
- 23968
- Gene Name
- NLR family, pyrin domain containing 5
- Gene Aliases
- Mater, Nalp5, Op1, PAN11
- Chromosome Location
- Chr. 7: 23385897 - 23441921 on Build GRCm38
- UniGene ID
- Mm.333653
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF074018.1 |
- |
8, 9 |
2193 |
|
AK087774.1 |
- |
8, 9 |
2215 |
|
AY196361.1 |
- |
8, 9 |
2193 |
|
AY196362.1 |
- |
8, 9 |
2193 |
|
AY329484.1 |
- |
7, 8 |
2145 |
|
AY329485.1 |
- |
8, 9 |
2193 |
|
AY329486.1 |
- |
8, 9 |
2193 |
|
AY329487.1 |
- |
7, 8 |
2145 |
|
AY329489.1 |
- |
8, 9 |
2193 |
|
AY329490.1 |
- |
8, 9 |
2193 |
|
BC053384.1 |
- |
9, 10 |
2260 |
30049556 |
View Full Product Details