Gene Transcripts

Gene Symbol
Gltscr1
Entrez Gene ID
243842
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
Chromosome Location
Chr. 7: 15970671 - 16048082 on Build GRCm38
UniGene ID
Mm.440249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081418.1 NP_001074887.1 13, 14 3733
GenBank mRNA AK220278.1 - 7, 8 4406

View Full Product Details