Gene Transcripts

Gene Symbol
Lypla2
Entrez Gene ID
26394
Gene Name
lysophospholipase 2
Gene Aliases
LysoII
Chromosome Location
Chr. 4: 135968224 - 135972594 on Build GRCm38
UniGene ID
Mm.34302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011942.1 NP_036072.1 7 414
GenBank mRNA AB009653.1 - 7 414
AK003689.1 - 7 381
AK075590.1 - 7 406
AK089112.1 - 7 406
AK167478.1 - 7 401
BC068120.1 - 7 440 5714650

View Full Product Details