Gene Transcripts

Gene Symbol
Tspan32
Entrez Gene ID
27027
Gene Name
tetraspanin 32
Gene Aliases
AW208513, Art-1, BB235973, D7Wsu37e, Phemx, Tssc6
Chromosome Location
Chr. 7: 143004902 - 143019486 on Build GRCm38
UniGene ID
Mm.28172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128080.2 NP_001121552.1 2 191
NM_001128081.1 NP_001121553.1 1 306
NM_001128082.1 NP_001121554.1 1 306
NM_020286.3 NP_064682.1 1 306
GenBank mRNA AF175771.1 - 1 155
AF291425.1 - 1 274
AJ279791.1 - 1 266
AJ279792.1 - 1 266
AJ279793.1 - 1 266
AJ279794.1 - 1 266
AJ279795.1 - 1 266
AK013248.1 - 1 82
AK017989.1 - 1 276
AK041748.1 - 2 177
AK154694.1 - 1 327
AK171199.1 - 1 305
AY039000.1 - 2 199
BC051204.1 - 1 296
BC055858.1 - 1 280 4913671
BY208601.1 - 1 324

View Full Product Details