Gene Transcripts

Gene Symbol
Tor1b
Entrez Gene ID
30934
Gene Name
torsin family 1, member B
Gene Aliases
2610016F05Rik, DQ1, torsinB
Chromosome Location
Chr. 2: 30952938 - 30959015 on Build GRCm38
UniGene ID
Mm.249164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133673.3 NP_598434.2 2 299
GenBank mRNA AJ297743.1 - 2 329
AK017290.1 - 2 318
AK028859.1 - 2 113
AK149754.1 - 2 320
AK159843.1 - 2 298
AK169692.1 - 2 322
BC018456.1 - 2 298 3582246

View Full Product Details