Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301204.1 | NP_001288133.1 | 3 | 495 | |
NM_001301205.1 | NP_001288134.1 | 2 | 180 | ||
NM_176913.4 | NP_795887.2 | 3 | 610 | ||
GenBank mRNA | AF488552.1 | - | 3 | 610 | |
AK089165.1 | - | 1 | 1787 | ||
AK089244.1 | - | 3 | 481 | ||
AK155308.1 | - | 3 | 446 | ||
AK155417.1 | - | 3 | 450 | ||
AK170094.1 | - | 3 | 447 | ||
AK170970.1 | - | 3 | 447 | ||
AK171044.1 | - | 3 | 447 | ||
AK171122.1 | - | 3 | 447 | ||
AK171155.1 | - | 2 | 160 | ||
AK171196.1 | - | 3 | 447 | ||
AK171268.1 | - | 3 | 447 | ||
AK171503.1 | - | 3 | 447 | ||
AK171606.1 | - | 3 | 463 |