Gene Transcripts

Gene Symbol
Ppm1h
Entrez Gene ID
319468
Gene Name
protein phosphatase 1H (PP2C domain containing)
Gene Aliases
A430075L18Rik, ARHCL1, C030002B11Rik
Chromosome Location
Chr. 10: 122678106 - 122945795 on Build GRCm38
UniGene ID
Mm.489703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110218.1 NP_001103688.1 2 727
NM_176919.4 NP_795893.2 2 727
GenBank mRNA AK029461.1 - 2 289
AK040194.1 - 2 471
AK040207.1 - 2 712
AK043121.1 - 2 728
AK051434.1 - 2 685
AK077422.1 - 2 716
AK134804.1 - 2 707
AK150309.1 - 2 289
AK220260.1 - 2 684
BC052910.1 - 1 38

View Full Product Details