Gene Transcripts

Gene Symbol
Igf2bp2
Entrez Gene ID
319765
Gene Name
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases
C330012H03Rik, IMP-2, Imp2, Neilsen
Chromosome Location
Chr. 16: 22059009 - 22163299 on Build GRCm38
UniGene ID
Mm.489442
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183029.2 NP_898850.2 1, 2 523
GenBank mRNA AK049196.1 - 1, 2 220
AK077404.1 - 1, 2 202
AK159654.1 - 1, 2 258
AY531659.1 - 1, 2 187
BC023758.1 - 1, 2 221
BC054552.1 - 1, 2 243
CB290059.1 - 1, 2 523

View Full Product Details