Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177320.2 | NP_796294.2 | 10 | 1393 | |
GenBank mRNA | AK029201.1 | - | 8 | 1061 | |
AK089483.1 | - | 10 | 1398 | ||
AK149847.1 | - | 10 | 1361 | ||
AK154643.1 | - | 10 | 1372 | ||
AK161050.1 | - | 1 | 301 | ||
AK171279.1 | - | 10 | 1394 | ||
AY156924.1 | - | 9 | 1242 | ||
BC128076.1 | - | 9 | 1242 | 40060132 | |
BC128077.1 | - | 9 | 1242 | 40060133 |