Gene Transcripts

Gene Symbol
Tas2r130
Entrez Gene ID
387355
Gene Name
taste receptor, type 2, member 130
Gene Aliases
STC74, T2R30, Tas2r30, Tas2r7, mt2r42
Chromosome Location
Chr. 6: 131629892 - 131630830 on Build GRCm38
UniGene ID
Mm.377916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199156.1 NP_954607.1 1 790
GenBank mRNA BC119329.1 - 1 872

View Full Product Details