Gene Transcripts

Gene Symbol
Gnptab
Entrez Gene ID
432486
Gene Name
N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits
Gene Aliases
EG432486, mKIAA1208
Chromosome Location
Chr. 10: 88379137 - 88447330 on Build GRCm38
UniGene ID
Mm.335190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004164.2 NP_001004164.2 8 805
GenBank mRNA AK140867.1 - 7 909
AK173132.1 - 8 805
BC060638.1 - 9 913 6849405

View Full Product Details