Gene Transcripts

Gene Symbol
Slc2a7
Entrez Gene ID
435818
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 7
Gene Aliases
OTTMUSG00000010396
Chromosome Location
Chr. 4: 150148837 - 150168895 on Build GRCm38
UniGene ID
Mm.359648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085529.1 NP_001078998.1 9 1090

View Full Product Details