Gene Transcripts

Gene Symbol
Snx1
Entrez Gene ID
56440
Gene Name
sorting nexin 1
Gene Aliases
Chromosome Location
Chr. 9: 66088127 - 66124886 on Build GRCm38
UniGene ID
Mm.271891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019727.2 NP_062701.2 9 964
GenBank mRNA AB019214.1 - 9 904
AF154120.1 - 9 968
AK149850.1 - 9 876
AK154079.1 - 9 877
AK155746.1 - 9 852
AK161241.1 - 9 945
AK167177.1 - 9 919
AK167993.1 - 9 859
AK168742.1 - 9 957
AK177911.1 - 3 154
AK189960.1 - 3 154
AK192993.1 - 3 154
BC066044.1 - 9 921 6807363
BC066189.1 - 9 855 30471747

View Full Product Details