Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019861.1 | NP_063914.1 | 13, 14 | 1455 | |
GenBank mRNA | AF136280.1 | - | 13, 14 | 1356 | |
AF197480.1 | - | 13, 14 | 1400 | ||
AJ131851.1 | - | 13, 14 | 1332 | ||
AK017474.1 | - | 13, 14 | 1337 | ||
AK075862.1 | - | 13, 14 | 1348 | ||
BC004054.1 | - | 10, 11 | 926 | ||
BC051131.1 | - | 12, 13 | 1574 | ||
BC058758.1 | - | 13, 14 | 1346 | 6492834 |