Gene Transcripts

Gene Symbol
Gmnn
Entrez Gene ID
57441
Gene Name
geminin
Gene Aliases
AW546347, Gem
Chromosome Location
Chr. 13: 24751845 - 24761937 on Build GRCm38
UniGene ID
Mm.12239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020567.2 NP_065592.1 5 453
GenBank mRNA AF068780.1 - 5 600
AK131959.1 - 5 427

View Full Product Details