Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025709.2 | NP_079985.2 | 27 | 4856 | |
GenBank mRNA | AK003521.1 | - | 3 | 322 | |
AK014474.1 | - | 12 | 1722 | ||
AK088851.1 | - | 11 | 1419 | ||
AK129381.1 | - | 26 | 4451 | ||
BC043715.1 | - | 6 | 820 | ||
BC057164.1 | - | 12 | 1725 | ||
BC060123.1 | - | 27 | 4856 | 6408778 | |
EF155419.1 | - | 25 | 4274 |