Gene Transcripts

Gene Symbol
Etfdh
Entrez Gene ID
66841
Gene Name
electron transferring flavoprotein, dehydrogenase
Gene Aliases
0610010I20Rik, AV001013
Chromosome Location
Chr. 3: 79603788 - 79628767 on Build GRCm38
UniGene ID
Mm.28336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025794.2 NP_080070.2 7 976
GenBank mRNA AK002483.1 - 7 929
AK075673.1 - 7 927
AK141684.1 - 7 930
AK152624.1 - 7 945
AK169686.1 - 7 881
BC012522.1 - 7 925 2648103
BC057670.1 - 7 943 2810976

View Full Product Details