Gene Transcripts

Gene Symbol
Tcf25
Entrez Gene ID
66855
Gene Name
transcription factor 25 (basic helix-loop-helix)
Gene Aliases
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location
Chr. 8: 123373635 - 123404174 on Build GRCm38
UniGene ID
Mm.178818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037877.3 NP_001032966.1 14, 15 1788
NM_001037878.3 NP_001032967.1 14, 15 1788
NM_001286362.1 NP_001273291.1 13, 14 1713
NM_001286363.1 NP_001273292.1 13, 14 1713
NM_025804.3 NP_080080.2 14, 15 1788
GenBank mRNA AK003180.1 - 14, 15 1675
AK013754.1 - 14, 15 1477
AK045397.1 - 13, 14 1641
AK050274.1 - 14, 15 1714
AK075728.1 - 14, 15 1638
AK076151.1 - 8, 9 875
AK088360.1 - 14, 15 1676
AK088602.1 - 1, 2 327
AK089964.1 - 14, 15 1677
AK122430.1 - 14, 15 1643
AK150918.1 - 14, 15 1669
AK150942.1 - 14, 15 1662
AK151143.1 - 14, 15 1662
AK151558.1 - 14, 15 1668
AK152178.1 - 14, 15 1662
AK152494.1 - 14, 15 1662
AK161627.1 - 14, 15 1664
AK167837.1 - 14, 15 1662
AK168167.1 - 14, 15 1662
BC016130.1 - 14, 15 1650
BC046768.1 - 14, 15 1746 5707312
EU624323.1 - 13, 14 1551
U94988.1 - 14, 15 1649

View Full Product Details