Gene Transcripts

Gene Symbol
Pof1b
Entrez Gene ID
69693
Gene Name
premature ovarian failure 1B
Gene Aliases
2310066B14Rik
Chromosome Location
Chr. X: 112638427 - 112698711 on Build GRCm38
UniGene ID
Mm.34964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181579.1 NP_853557.1 13 1490
GenBank mRNA AF309775.1 - 12 1383
AK036922.1 - 13 1497
AK137653.1 - 13 1489

View Full Product Details