Gene Transcripts

Gene Symbol
Ipmk
Entrez Gene ID
69718
Gene Name
inositol polyphosphate multikinase
Gene Aliases
2410017C19Rik, AA408208, Impk
Chromosome Location
Chr. 10: 71347765 - 71385885 on Build GRCm38
UniGene ID
Mm.245867
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027184.1 NP_081460.1 2 318
GenBank mRNA AK010523.1 - 2 277
AK036084.1 - 2 86
AK172246.1 - 2 276
BC052463.1 - 2 318 6834748

View Full Product Details